Learn More
Abcam Mouse Fibrinogen ELISA Kit- Intracellular, 1 x 96 Tests
Catalog No. 502698084
Encompass_Preferred
Mouse Fibrinogen ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Mouse Fibrinogen with a sensitivity of 67 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8084 Supplier Abcam Supplier No. AB3157891X96TESTS
Mouse Fibrinogen ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Mouse Fibrinogen with a sensitivity of 67 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement