Learn More
Biomatik Corporation Mouse Fibrinogen Gamma (FGg) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11639
Encompass_Preferred
Mouse Fibrinogen Gamma (FGg) Mini sample ELISA Kit for detecting FGg in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.4ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VCM7. Gene ID: 99571. Target Synonyms: FG-G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8112