Learn More
Innovative Research Inc Mouse Fibrinogen Purified 1mg
Supplier:  Innovative Research Inc IMSFBG1MG
Encompass_Preferred
Mouse Fibrinogen Purified from Innovative Research has been purified from plasminogen depleted Mouse Plasma using a combination of several chromatographic and fractionation steps. This is a frozen liquid buffered in 0.02M Sodium Citrate-HCl; pH 7.4, with a concentration of 2.0 mg/ml and a purity of >95%.
Catalog No. 50-203-6456