Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 13 (FGF13) CLIA Kit
Supplier:  Biomatik Corporation EKU09803
Encompass_Preferred
Mouse Fibroblast Growth Factor 13 (FGF13) CLIA Kit for detecting FGF13 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.98pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P70377. Gene ID: 14168. Target Synonyms: FHF2; Fibroblast Growth Factor Homologous Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6556