Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 5 (FGF5) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11122
Encompass_Preferred
Mouse Fibroblast Growth Factor 5 (FGF5) Mini sample ELISA Kit for detecting FGF5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15656. Gene ID: 14176. Target Synonyms: HBGF5; Smag82; Heparin-binding growth factor 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6600