Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 18 (FGF18) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11113
Encompass_Preferred
Mouse Fibroblast Growth Factor 18 (FGF18) Mini sample ELISA Kit for detecting FGF18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O89101. Gene ID: 14172. Target Synonyms: ZFGF5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8219