Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 3 (FGF3) CLIA Kit
Supplier:  Biomatik Corporation EKU09817
Encompass_Preferred
Mouse Fibroblast Growth Factor 3 (FGF3) CLIA Kit for detecting FGF3 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.57pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05524. Target Synonyms: HBGF3; INT2; Heparin-binding growth factor 3; Murine Mammary Tumor Virus Integration Site (v-int-2) Oncogene Homolog; INT-2 Proto-Oncogene Protein; Proto-oncogene Int-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8429