Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 12 (FGF12) CLIA Kit
Supplier:  Biomatik Corporation EKU09801
Encompass_Preferred
Mouse Fibroblast Growth Factor 12 (FGF12) CLIA Kit for detecting FGF12 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P61329-2. Gene ID: 14167. Target Synonyms: FGF12B; FHF1; Fibroblast Growth Factor Homologous Factor 1; Myocyte-Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8431