Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 20 (FGF20) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11116
Encompass_Preferred
Mouse Fibroblast Growth Factor 20 (FGF20) Mini sample ELISA Kit for detecting FGF20 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESL9. Gene ID: 80857. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8218