Learn More
American Research Products Inc Mouse fibroblast growth factor
Catalog No. 5000329992
Encompass
Mouse fibroblast growth factor 10 (FGF10) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29992 Supplier American Research Products Inc Supplier No. CSBE13045M
Mouse fibroblast growth factor 10 (FGF10) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich