Learn More
American Research Products Inc Mouse Fibroblast growth factor
Supplier:  American Research Products Inc CSBEL008629MO
Encompass
Mouse Fibroblast growth factor 23 (FGF23) ELISA kit from CUSABIO Detection Range 23 44 pg/mL-1500 pg/mL Sensitivity 5 86 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26923