Learn More
American Research Products Inc Mouse Fibroblast growth factor
Supplier:  American Research Products Inc CSBEL008627MO
Encompass
Mouse Fibroblast growth factor 21 (FGF21) ELISA kit from CUSABIO Detection Range 31 25 pg/ml -2000 pg/ml Sensitivity 7 81 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27686