Learn More
American Research Products Inc Mouse Fibroblast growth factor
Supplier:  American Research Products Inc CSBEL008632MO
Encompass
Mouse Fibroblast growth factor 5 (FGF5) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26891