Learn More
American Research Products Inc Mouse Fibroblast growth factor
Catalog No. 5000327242
Encompass
Mouse Fibroblast growth factor 15 (FGF15) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27242 Supplier American Research Products Inc Supplier No. CSBEL522052MO
Mouse Fibroblast growth factor 15 (FGF15) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates cell lysates Method Sandwich