Learn More
Biomatik Corporation Mouse Ficolin 2 (FCN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11704
Encompass_Preferred
Mouse Ficolin 2 (FCN2) ELISA Kit for detecting FCN2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.247ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70497. Gene ID: 14134. Target Synonyms: EBP37; FCNL; P35; Hucolin; 37 kDa elastin-binding protein; Collagen/Fibrinogen Domain Containing Lectin; Ficolin B; Serum Lectin p35; L-Ficolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6535