Learn More
Cusabio Technology LLC Mouse Ficolin-2 FCN2 ELISA kit
Catalog No. 5000049116
Encompass_Preferred
The Mouse Ficolin-2 (FCN2) ELISA Kit is designed for the quantitative detection of FCN2 in serum plasma and tissue homogenates using a sandwich assay format With a detection range of 125 pg/mL to 8000 pg/mL and a sensitivity of 31 25 pg/mL this kit provides precise measurement of FCN2 concentrations in biological samples The assay requires a sample volume of 50-100 L and employs a 450 nm wavelength for absorbance readings The protocol involves an incubation period of 1-5 hours balancing efficiency with accuracy This kit is suitable for researchers studying immune-related pathways as FCN2 plays a role in innate immunity through its lectin-based complement activation The standardized components ensure reproducibility across experimental setups
Catalog No. 50-000-49116 Supplier Cusabio Technology LLC Supplier No. CSBEL008551MO
The Mouse Ficolin-2 (FCN2) ELISA Kit is designed for the quantitative detection of FCN2 in serum plasma and tissue homogenates using a sandwich assay format With a detection range of 125 pg/mL to 8000 pg/mL and a sensitivity of 31 25 pg/mL this kit provides precise measurement of FCN2 concentrations in biological samples The assay requires a sample volume of 50-100 L and employs a 450 nm wavelength for absorbance readings The protocol involves an incubation period of 1-5 hours balancing efficiency with accuracy This kit is suitable for researchers studying immune-related pathways as FCN2 plays a role in innate immunity through its lectin-based complement activation The standardized components ensure reproducibility across experimental setups