Learn More
Biomatik Corporation Mouse FK506 Binding Protein Like Protein (FKBPL) ELISA Kit
Supplier:  Biomatik Corporation EKU04235
Encompass_Preferred
Mouse FK506 Binding Protein Like Protein (FKBPL) ELISA Kit for detecting FKBPL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35450. Gene ID: 56299. Target Synonyms: NG7; WISP39; DIR1; WAF-1/CIP1 stabilizing protein 39. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5458