Learn More
BIOHIPPO Mouse FKBPL Microsample
SDP
Catalog No. 5000314645
Encompass
Mouse FKBPL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FKBPL in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 111 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FKBPL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FKBPL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FKBPL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14645 Supplier BIOHIPPO Supplier No. BHE15210733
Mouse FKBPL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FKBPL in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 111 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FKBPL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FKBPL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FKBPL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color