Learn More
Sino Biological Recombinant Mouse VEGFR1/FLT1 Protein (ECD, His Tag)
Supplier:  Sino Biological 51045M08H500
Encompass_Preferred
A DNA sequence encoding the mouse FLT1 (NP_034358.2) (Met1-Glu759) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3020