Learn More
FUJIFILM Biosciences Mouse Flt3-Ligand , Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50990809
Encompass_Preferred
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation.  FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor.  FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).  
Catalog No. 50-990-809 Supplier FUJIFILM Biosciences Supplier No. 20006100UG
Add to Cart
Edge
Add to Cart
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation.  FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor.  FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).  