Learn More
FUJIFILM BIOSCIENCES INC Mouse Flt3-Ligand , Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2000610UG
Encompass_Preferred
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation.  FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor.  FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).  
Catalog No. 50-990-808