Learn More
Sino Biological Recombinant Mouse Flt3 Ligand/FLT3LG Protein (hFc Tag)
Supplier:  Sino Biological 51113M02H1
Encompass_Preferred
A DNA sequence encoding the mouse FLT3L (P49772-1) (Met1-Arg188) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3001