Learn More
Sino Biological Recombinant Anti-VEGFR3/FLT4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50584R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VEGFR3 / FLT-4 (rM VEGFR3 / FLT-4; Catalog#50584-M08H; P35917-1; Met1-Glu775).
Catalog No. 50-240-8203