Learn More
Sino Biological Anti-VEGFR3/FLT4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50584RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse VEGFR3 / FLT-4 (rM VEGFR3 / FLT-4; Catalog#50584-M08H; P35917-1; Met 1-Glu 775). VEGFR3 / FLT-4 specific IgG was purified by Mouse VEGFR3 / FLT-4 affinity chromatography.
Catalog No. 50-241-2572