Learn More
ACROBiosystems Alexa Fluor 647-Labeled Monoclonal Anti-FMC63 Antibody, Mouse IgG1 (Y45)Star Staining
Supplier:  ACROBiosystems FM3AM534200TESTS
Encompass_Preferred
Alexa Fluor 647-Labeled Monoclonal Anti-FMC63 Antibody, Mouse IgG1 (Y45) (FM3-AM534) is produced via conjugation of AF647 to Monoclonal Anti-FMC63 Antibody, Mouse IgG1 under optimal conditions with a new generation site-specific technology under Star Staining labeling platform.
Catalog No. 50-233-1258