Learn More
ACROBiosystems FITC-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is expressed from human HEK293 cells. It is the FITC labeled form of Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-Y45P1).
Catalog No. 502231410
Encompass_Preferred
FMC63 is an IgG2a mouse monoclonal antibody specific for CD19, which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. So far, most of reported CART19 trials contain the anti-CD19 scFv derived from FMC63, including the two FDA-approved CARs Kymriah and Yescarta.
This item is not returnable. View return policy
Catalog No. 50-223-1410 Supplier ACROBiosystems Supplier No. FM3FY45P125TESTS
Add to Cart
Add to Cart
FMC63 is an IgG2a mouse monoclonal antibody specific for CD19, which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. So far, most of reported CART19 trials contain the anti-CD19 scFv derived from FMC63, including the two FDA-approved CARs Kymriah and Yescarta.