Learn More
ACROBiosystems Biotinylated FMC63 scFv, Fc,Avitag™ (MALS verified)
Catalog No. 502331850
Encompass_Preferred
Biotinylated FMC63 scFv, Fc,Avitag is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293), which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63.
Catalog No. 50-233-1850 Supplier ACROBiosystems Supplier No. CD9BV4324B25UG
Biotinylated FMC63 scFv, Fc,Avitag is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293), which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63.