Learn More
Biomatik Corporation Mouse FMS Like Tyrosine Kinase 3 Ligand (Flt3L) CLIA Kit
Supplier:  Biomatik Corporation EKU09730
Encompass_Preferred
Mouse FMS Like Tyrosine Kinase 3 Ligand (Flt3L) CLIA Kit for detecting Flt3L in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P49772. Gene ID: 14256. Target Synonyms: FLT3LG; FLT3-LG; Flt3-L; FL; Fms-Related Tyrosine Kinase 3 ligand; SL cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5576