Learn More
Biomatik Corporation Mouse Folic Acid, Fa Elisa Kit
Supplier:  Biomatik Corporation EKC31394
Encompass_Preferred
Mouse Folic Acid, Fa Elisa Kit for detecting Folic Acid, Fa in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.195-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9189