Learn More
Biomatik Corporation Mouse Follicle Stimulating Hormone (FSH) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12306
Encompass_Preferred
Mouse Follicle Stimulating Hormone (FSH) Mini sample ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.88ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01216. Gene ID: 12640. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7987