Learn More
Biomatik Corporation Mouse Follicle Stimulating Hormone Beta (FSHb) ELISA Kit
Supplier:  Biomatik Corporation EKU10844
Encompass_Preferred
Mouse Follicle Stimulating Hormone Beta (FSHb) ELISA Kit for detecting FSHb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.94-60ng/ml. Sensitivity: 0.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60687. Gene ID: 14308. Target Synonyms: FSH-B; Follitropin Subunit Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7513