Learn More
Sino Biological Recombinant Mouse FOLR1 Protein (His Tag) 1
Supplier:  Sino Biological 50573M08H1
Encompass_Preferred
A DNA sequence encoding the mouse FOLR1 (NP_032060.2) (Met 1-Met 231) without the pro peptide was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7251