Learn More
Sino Biological Anti-FOLR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50573T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FOLR1 (rM FOLR1; Catalog#50573-M08H; NP_032060.2; Met1-Met231). FOLR1 specific IgG was purified by Mouse FOLR1 affinity chromatography.
Catalog No. 50-255-3213