Learn More
ACROBiosystems Mouse FOLR1 Protein Fc Tag
Supplier:  ACROBiosystems FO1M5254100UG
Encompass_Preferred
Mouse FOLR1 Protein Fc Tag (FO1-M5254) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).
Catalog No. 50-253-5050