Learn More
Biomatik Corporation Mouse Forkhead Box Protein P1 (FOXP1) ELISA Kit
Supplier:  Biomatik Corporation EKU10108
Encompass_Preferred
Mouse Forkhead Box Protein P1 (FOXP1) ELISA Kit for detecting FOXP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58462. Gene ID: 108655. Target Synonyms: FOX-P1; 12CC4; QRF1; HFKH1B; Fork Head-Related Protein Like B; Glutamine-Rich Factor 1; PAX5/FOXP1 Fusion Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8368