Learn More
Abcam Mouse FOXP3 ELISA Kit, 1 x 96 Tests
Catalog No. 502654600
Encompass_Preferred
Mouse FOXP3 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse FOXP3 with a sensitivity of 0.3 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4600 Supplier Abcam Supplier No. AB2896451X96TESTS
Mouse FOXP3 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse FOXP3 with a sensitivity of 0.3 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement