Learn More
Raybiotech Inc MOUSE FRACTALKINE ELISA
Supplier:  Raybiotech Inc ELMFRACTALKINE1
Encompass_Preferred
RayBio Mouse Fractalkine (CX3CL1) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 0.3 ng/ml - 200 ng/ml
Catalog No. 50-104-4653