Learn More
American Research Products Inc Mouse free testosterone F TES
Catalog No. 5000329382
Encompass
Mouse free testosterone (F-TESTO) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 4 pg/ml Sample serum plasma Method Competitive
Catalog No. 50-003-29382 Supplier American Research Products Inc Supplier No. CSBE05098M
Mouse free testosterone (F-TESTO) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 4 pg/ml Sample serum plasma Method Competitive