Learn More
American Research Products Inc Mouse Free ThyroXine FT4 ELIS
Supplier:  American Research Products Inc CSBE05080M
Encompass
Mouse Free ThyroXine FT4 ELISA Kit from CUSABIO Detection Range 4 pmol/L-64 pmol/L Sensitivity 1 08 pmol/L Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26827