Learn More
Biomatik Corporation Mouse Free Thyroxine, Ft4 Elisa Kit
Supplier:  Biomatik Corporation EKC31396
Encompass_Preferred
Mouse Free Thyroxine, Ft4 Elisa Kit for detecting Free Thyroxine, Ft4 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4-64pmol/l. Sensitivity: 1.08pmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9188