Learn More
Biomatik Corporation Mouse Free Tri-Iodothyronine, Free-T3 Elisa Kit
Supplier:  Biomatik Corporation EKC31397
Encompass_Preferred
Mouse Free Tri-Iodothyronine, Free-T3 Elisa Kit for detecting Free Tri-Iodothyronine Indes, Free-T3 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 2-32pmol/l. Sensitivity: 0.38pmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9187