Learn More
Sino Biological Anti-Follistatin-like 1/FSTL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51127T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse FSTL1 (Catalog#51127-M08H; NP_032073.2; Met1-Ile306). FSTL1 specific IgG was purified by Mouse FSTL1 affinity chromatography.
Catalog No. 50-255-2990