Learn More
BIOHIPPO Mouse FTO Microsample
SDP
Catalog No. 5000315371
Encompass
Mouse FTO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FTO in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FTO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FTO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FTO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15371 Supplier BIOHIPPO Supplier No. BHE15210626
Mouse FTO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FTO in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse FTO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse FTO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse FTO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color