Learn More
Biomatik Corporation Mouse Fucosyltransferase 8 (FUT8) ELISA Kit
Supplier:  Biomatik Corporation EKU04286
Encompass_Preferred
Mouse Fucosyltransferase 8 (FUT8) ELISA Kit for detecting FUT8 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTS2. Gene ID: 53618. Target Synonyms: GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase; Glycoprotein 6-alpha-L-fucosyltransferase; Alpha-(1,6)-fucosyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6032