Learn More
Abcam Mouse G-CSF Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502672509
Encompass_Preferred
Mouse G-CSF Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse G-CSF.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2509 Supplier Abcam Supplier No. AB24168210X96TESTS
Mouse G-CSF Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse G-CSF.
The product is subject to the following: Abcam Restricted Use Statement