Learn More
FUJIFILM BIOSCIENCES INC Mouse G-CSF, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200141MG
Encompass_Preferred
Granulocyte-colony stimulating factor (G-CSF) is a cytokine that functions as a potent inducer of neutrophilic granulocyte proliferation, terminal differentiation, and activation.  G-CSF synthesis occurs in monocyte, macrophage, epithelial, endothelial, and fibroblast cells after activation by bacterial endotoxins, tumor necrosis factor alpha (TNF-A), interleukin 1 (IL-1), or interleukin 17 (IL-17).  The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways.  G-CSF also promotes neurogenesis and inhibits neuronal apoptosis.  Human and mouse G-CSF proteins are cross-reactive. 
Catalog No. 50-990-837