Learn More
R&D Systems™ Mouse G6b (NP_001028393) VersaClone cDNA
Description
- Mpig6b megakaryocyte and platelet inhibitory receptor G6b [ Mus musculus (house mouse) ]
- Also known as: G6b; Ng31; C6orf25; AU023871
- The G6b/C6orf25 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- G6B is a member of the immunoglobulin (Ig) superfamily and is located in the major histocompatibility complex (MHC) class III region.
- It is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms have been found in the endoplasmic reticulum and Golgi before being secreted.
- It is expressed mainly on the platelet lineage. Crosslinking of G6b inhibits agonist-stimulated platelet aggregation and activation.
- Heparin interacts with the G6b extracellular domain with high-affinity.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mMpig6b |
| Accession Number | NP_001028393 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 607 bp |
| Gene ID (Entrez) | 80739 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.