Learn More
Cusabio Technology LLC MOUSE GAD au-Ab IgG ELISA KIT
Catalog No. 5000049673
Encompass_Preferred
This ELISA kit detects GAD au-Ab (IgG) in serum and plasma samples using an indirect qualitative method Designed for efficient analysis the assay requires a sample volume of 50-100 L and delivers results within 1-5 hours The detection is performed by measuring absorbance at 450 nm ensuring reliable identification of glutamic acid decarboxylase autoantibodies of the IgG class This kit is optimized for specificity and sensitivity providing a standardized tool for research applications requiring the assessment of autoimmune responses associated with GAD antibodies Suitable for laboratory use it offers a straightforward protocol compatible with standard ELISA equipment and workflows
Catalog No. 50-000-49673 Supplier Cusabio Technology LLC Supplier No. CSBE08655M
This ELISA kit detects GAD au-Ab (IgG) in serum and plasma samples using an indirect qualitative method Designed for efficient analysis the assay requires a sample volume of 50-100 L and delivers results within 1-5 hours The detection is performed by measuring absorbance at 450 nm ensuring reliable identification of glutamic acid decarboxylase autoantibodies of the IgG class This kit is optimized for specificity and sensitivity providing a standardized tool for research applications requiring the assessment of autoimmune responses associated with GAD antibodies Suitable for laboratory use it offers a straightforward protocol compatible with standard ELISA equipment and workflows