Learn More
BIOHIPPO Mouse GAL12 Microsample
SDP
Catalog No. 5000315302
Encompass
Mouse GAL12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL12 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GAL12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GAL12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GAL12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15302 Supplier BIOHIPPO Supplier No. BHE15211873
Mouse GAL12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GAL12 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GAL12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GAL12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GAL12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color