Learn More
American Research Products Inc Mouse galanin prepropeptide G
Supplier:  American Research Products Inc CSBEL009191MO
Encompass
Mouse galanin prepropeptide (GAL) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27806